missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Methylbenzyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65426.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15412559

  • £119.78 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Methylbenzyl)-2,2-diphenylacetamide
315.42
MFCD01345955
N-(p-Tolylmethanamine)-2,2-diphenylacetamide

Specifications

N-(4-Methylbenzyl)-2,2-diphenylacetamide
Solid
315.42
N-(p-Tolylmethanamine)-2,2-diphenylacetamide
1g
95%
MFCD01345955
Videos
SDS