missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Methoxyphenyl)-p-phenylenediamine Hydrochloride 95%, ACROS Organics™

Brand:  Acros Organics 413890250

Code : 29

Additional Details : CAS Number : 3566-44-7 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10617001

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

N-(4-Methoxyphenyl)-p-phenylenediamine hydrochloride
3566-44-7
100.0
95%
250.73
95%
94.0
95%
C13H14N2O·HCl
MFCD00012989

Specifications

N-(4-Methoxyphenyl)-p-phenylenediamine hydrochloride
247.0°C
95%
94.0
Authentic
Crystalline Powder
95%
250.73
13, III, 1161
Gray-blue
25g
3566-44-7
100.0
Glass bottle
95%
C13H14N2O·HCl
MFCD00012989
15, 6072
Videos
SDS