missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Methoxyphenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65879.03

Code : NEW

Additional Details : CAS Number : 23105-44-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15422659

  • £85.44 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Methoxyphenyl)-2,2-diphenylacetamide
317.39
2662309
23105-44-4
MFCD00039803

Specifications

N-(4-Methoxyphenyl)-2,2-diphenylacetamide
23105-44-4
95%
MFCD00039803
1g
Solid
317.39
2662309
Videos
SDS