Alfa Aesar™ N-(4-Methoxybenzyl)-2-nitrobenzenesulfonamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H60564.03

1GR N-(4-Methoxybenzyl)-2-nitrobenzenesulfonamide, 97% 1g

UNSPSC: 12352100

Code: NEW

Additional Details:
Additional Details: CAS Number: 171414-16-7 Weight: 0.00100kg


Quantity 1g

Product Code. 15467048

Quantity Price
1 £ 358.49 / 1g
Estimated Shipment date
from Supplier 07-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-(4-Methoxybenzyl)-2-nitrobenzenesulfonamide
CAS 171414-16-7
Formula Weight 322.34
MDL Number MFCD01212786
Beilstein 7307319

Specification

Formula Weight 322.34
Chemical Name or Material N-(4-Methoxybenzyl)-2-nitrobenzenesulfonamide
Physical Form Solid
Assay 97%
MDL Number MFCD01212786
CAS 171414-16-7
Beilstein 7307319
Quantity 1g