missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Isopropylphenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65259.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15422499

  • £91.90 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Isopropylphenyl)-2,2-diphenylacetamide
329.44
MFCD00588525

Specifications

N-(4-Isopropylphenyl)-2,2-diphenylacetamide
Solid
329.44
1g
95%
MFCD00588525
Videos
SDS