missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Iodophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60164.03

Code : NEW

Additional Details : CAS Number : 21226-24-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15446638

  • £358.49 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Iodophenyl)benzenesulfonamide
21226-24-4
359.18
MFCD00441555

Specifications

N-(4-Iodophenyl)benzenesulfonamide
White to pale orange
21226-24-4
97%
MFCD00441555
Light Sensitive
1g
Solid
359.18
Videos
SDS