missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Hydroxyphenyl)glycine 99%, ACROS Organics™

Brand:  Acros Organics 121730250

Code : 29

Additional Details : CAS Number : 122-87-2 Weight : 0.07500kg


Quantity 25g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10647272

  • £24.30 / 25g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

N-(4-Hydroxyphenyl)glycine
122-87-2
100.0
99%
HOC6H4NHCH2CO2H
MFCD00002689
99%
98.5
99%
C8H9NO3
167.16
Glycin

Specifications

N-(4-Hydroxyphenyl)glycine
248.0°C
99%
98.5
Authentic
Plastic bottle
99%
C8H9NO3
167.16
13, 488
Glycin
White to beige-brown or brown-gray
25g
122-87-2
100.0
1% max.
Powder
99%
HOC6H4NHCH2CO2H
MFCD00002689
15, 4875
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N-(4-Hydroxyphenyl)glycine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS