missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Hydroxyphenyl)glycine 99%, ACROS Organics™

Brand:  Acros Organics 121731000

Code : 29

Additional Details : CAS Number : 122-87-2 Weight : 0.15000kg


Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10162200

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

N-(4-Hydroxyphenyl)glycine
122-87-2
100.0
99%
HOC6H4NHCH2CO2H
MFCD00002689
99%
98.5
99%
C8H9NO3
167.16
Glycin

Specifications

N-(4-Hydroxyphenyl)glycine
248.0°C
99%
98.5
Authentic
Plastic bottle
99%
C8H9NO3
167.16
13, 488
Glycin
White to beige-brown or brown-gray
100g
122-87-2
100.0
1% max.
Powder
99%
HOC6H4NHCH2CO2H
MFCD00002689
15, 4875
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N-(4-Hydroxyphenyl)glycine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS