missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Fluorobenzyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59824.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416248

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Fluorobenzyl)-3,5-dinitrobenzamide
319.24
MFCD00751050

Specifications

N-(4-Fluorobenzyl)-3,5-dinitrobenzamide
97%
MFCD00751050
Solid
319.24
1g
Videos
SDS