missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[4-Fluoro-2-(trifluoromethyl)phenyl]-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65363.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499586

  • £59.78 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-[4-Fluoro-2-(trifluoromethyl)phenyl]-2,2-diphenylacetamide
373.35
MFCD27975752

Specifications

N-[4-Fluoro-2-(trifluoromethyl)phenyl]-2,2-diphenylacetamide
95%
MFCD27975752
Solid
373.35
1g
Videos
SDS