missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Fluoro-2-methylphenyl)-2-methoxybenzamide, 97%

Brand:  Alfa Aesar™ H59012.03

Code : NEW

Additional Details : CAS Number : 895666-55-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15454716

  • £358.49 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Fluoro-2-methylphenyl)-2-methoxybenzamide
895666-55-4
259.28
MFCD07264152

Specifications

N-(4-Fluoro-2-methylphenyl)-2-methoxybenzamide
Solid
259.28
1g
895666-55-4
97%
MFCD07264152
Videos
SDS