missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Ethoxyphenyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59751.03

Code : NEW

Additional Details : CAS Number : 36293-20-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15466148

  • £358.49 / 1g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Ethoxyphenyl)-3,5-dinitrobenzamide
331.28
2900903
36293-20-6
MFCD00383713

Specifications

N-(4-Ethoxyphenyl)-3,5-dinitrobenzamide
Solid
331.28
2900903
1g
36293-20-6
97%
MFCD00383713
Yellow to pale brown
Videos
SDS