missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chlorophenyl)thiourea, 97%, Thermo Scientific™

Catalog Number 10057822
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 3696-23-9
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H7ClN2S
Linear Formula ClC6H4NHCSNH2
MDL Number MFCD00022168
Quantity 1 g
Synonym 1-4-chlorophenyl thiourea, 4-chlorophenyl thiourea, n-4-chlorophenyl thiourea, thiourea, 4-chlorophenyl, 1-p-chlorophenyl thiourea, n-p-chlorophenyl thiourea, 1-p-chlorophenyl-2-thiourea, 4-chlorophenyl-2-thiourea, urea, 1-p-chlorophenyl-2-thio
InChI Key XVEFWRUIYOXUGG-UHFFFAOYSA-N
SMILES NC(=S)NC1=CC=C(Cl)C=C1
IUPAC Name (4-chlorophenyl)thiourea
Molecular Weight (g/mol) 186.66
PubChem CID 735843
Formula Weight 186.67
Percent Purity 97%
Chemical Name or Material N-(4-chlorophenyl)thiourea
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'