missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chlorophenyl)benzenesulfonamide, 97%, Thermo Scientific™

Catalog Number 15496498
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
4750-28-1
C12H10ClNO2S
267.727
MFCD00541864
ANRCRHLXUCJAKV-UHFFFAOYSA-N
n-4-chlorophenyl benzenesulfonamide, benzenesulfonamide, n-4-chlorophenyl, benzenesulfonamide,n-4-chlorophenyl, p-chlorobenzenesulfonanilide, benzenesulfonanilide, 4'-chloro, n-4-chlorophenyl benzenesulfonamide #
20865
N-(4-chlorophenyl)benzenesulfonamide
C1=CC=C(C=C1)S(=O)(=O)NC2=CC=C(C=C2)Cl
This item is not returnable. View return policy
4750-28-1
C12H10ClNO2S
267.727
MFCD00541864
ANRCRHLXUCJAKV-UHFFFAOYSA-N
n-4-chlorophenyl benzenesulfonamide, benzenesulfonamide, n-4-chlorophenyl, benzenesulfonamide,n-4-chlorophenyl, p-chlorobenzenesulfonanilide, benzenesulfonanilide, 4'-chloro, n-4-chlorophenyl benzenesulfonamide #
20865
N-(4-chlorophenyl)benzenesulfonamide
C1=CC=C(C=C1)S(=O)(=O)NC2=CC=C(C=C2)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Purple
Quantity 250 mg
Formula Weight 267.73
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-(4-Chlorophenyl)benzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'