missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Chlorophenyl)-3,4-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61733.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15436746

  • £59.78 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Chlorophenyl)-3,4-difluorobenzenesulfonamide
303.71
MFCD06022707

Specifications

N-(4-Chlorophenyl)-3,4-difluorobenzenesulfonamide
97%
MFCD06022707
Solid
303.71
1g
Videos
SDS