missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Chlorophenyl)-2-nitrobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60800.03

Code : NEW

Additional Details : CAS Number : 63228-67-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447298

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Chlorophenyl)-2-nitrobenzenesulfonamide
63228-67-1
312.73
MFCD00577611

Specifications

N-(4-Chlorophenyl)-2-nitrobenzenesulfonamide
Solid
312.73
1g
63228-67-1
97%
MFCD00577611
Videos
SDS