missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Chlorobenzyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65611.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402589

  • £119.78 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Chlorobenzyl)-2,2-diphenylacetamide
335.83
MFCD01060909

Specifications

N-(4-Chlorobenzyl)-2,2-diphenylacetamide
95%
MFCD01060909
Solid
335.83
1g
Videos
SDS