missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[4-Chloro-3-(trifluoromethyl)phenyl]-3,4-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61132.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15426296

  • £63.71 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

N-[4-Chloro-3-(trifluoromethyl)phenyl]-3,4-difluorobenzenesulfonamide
371.71
MFCD05145171

Specifications

N-[4-Chloro-3-(trifluoromethyl)phenyl]-3,4-difluorobenzenesulfonamide
97%
MFCD05145171
Solid
371.71
1g
Videos
SDS