missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[4-Chloro-3-(trifluoromethyl)phenyl]-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65650.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429806

  • £74.73 / 1g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

N-[4-Chloro-3-(trifluoromethyl)phenyl]-2,2-diphenylacetamide
389.8
MFCD01230726

Specifications

N-[4-Chloro-3-(trifluoromethyl)phenyl]-2,2-diphenylacetamide
95%
MFCD01230726
Solid
389.8
1g
Videos
SDS