missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Chloro-3-methylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65978.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15400067

  • £34.13 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Chloro-3-methylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
299.8
MFCD27975849

Specifications

N-(4-Chloro-3-methylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
95%
MFCD27975849
Solid
299.8
1g
Videos
SDS