missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N-(4-Chloro-3-methylphenyl)-3,5-difluorobenzenesulfonamide, 97%
Brand: Alfa Aesar™ H62950.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
N-(4-Chloro-3-methylphenyl)-3,5-difluorobenzenesulfonamide | |
317.74 | |
MFCD26143349 |
Specifications
N-(4-Chloro-3-methylphenyl)-3,5-difluorobenzenesulfonamide | |
97% | |
MFCD26143349 |
Solid | |
317.74 | |
1g |