missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Chloro-2-methylphenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59718.03

Code : NEW

Additional Details : CAS Number : 349090-12-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15426118

  • £358.49 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Chloro-2-methylphenyl)-3-methylbenzamide
349090-12-6
259.74
MFCD00783697

Specifications

N-(4-Chloro-2-methylphenyl)-3-methylbenzamide
Solid
259.74
1g
349090-12-6
97%
MFCD00783697
Videos
SDS