missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Chloro-2-methylphenyl)-2-fluorobenzamide, 97%

Brand:  Alfa Aesar™ H59849.03

Code : NEW

Additional Details : CAS Number : 459130-69-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15436278

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Chloro-2-methylphenyl)-2-fluorobenzamide
459130-69-9
263.7
MFCD02068850

Specifications

N-(4-Chloro-2-methylphenyl)-2-fluorobenzamide
Solid
263.7
1g
459130-69-9
97%
MFCD02068850
Videos
SDS