missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Chloro-2-fluorophenyl)-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62684.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15487726

  • £68.26 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Chloro-2-fluorophenyl)-3,5-difluorobenzenesulfonamide
321.7
MFCD26143334

Specifications

N-(4-Chloro-2-fluorophenyl)-3,5-difluorobenzenesulfonamide
97%
MFCD26143334
Solid
321.7
1g
Videos
SDS