missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Chloro-2-fluorophenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65629.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449786

  • £79.71 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Chloro-2-fluorophenyl)-2,2-diphenylacetamide
339.79
MFCD03393076

Specifications

N-(4-Chloro-2-fluorophenyl)-2,2-diphenylacetamide
95%
MFCD03393076
Solid
339.79
1g
Videos
SDS