missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65063.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419356

  • £74.73 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
330.23
MFCD28098745

Specifications

N-(4-Bromophenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
Solid
330.23
1g
95%
MFCD28098745
Videos
SDS