missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromophenyl)-3,5-difluorobenzamide, 97%

Brand:  Alfa Aesar™ H61600.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458158

  • £73.56 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromophenyl)-3,5-difluorobenzamide
312.11
MFCD04806221

Specifications

N-(4-Bromophenyl)-3,5-difluorobenzamide
Solid
312.11
1g
97%
MFCD04806221
Videos
SDS