Alfa Aesar™ N-(4-Bromophenyl)-2,2-diphenylacetamide, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H65961.MD

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: CAS Number: 70481-06-0 Weight: 0.00025kg


Quantity 250mg

Product Code. 15432679

Quantity Price
1 £ 19.93 / 250mg
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-(4-Bromophenyl)-2,2-diphenylacetamide
CAS 70481-06-0
Formula Weight 366.26
MDL Number MFCD00028013
Beilstein 2143809

Specification

Formula Weight 366.26
Assay 95%
CAS 70481-06-0
Quantity 250mg
Physical Form Solid
MDL Number MFCD00028013
Chemical Name or Material N-(4-Bromophenyl)-2,2-diphenylacetamide
Beilstein 2143809
Documentation

Technical documents