missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromobenzyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59058.03

Code : NEW

Additional Details : CAS Number : 1291773-90-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475238

  • £358.49 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromobenzyl)-3-methylbenzamide
1291773-90-4
304.19
MFCD13481969

Specifications

N-(4-Bromobenzyl)-3-methylbenzamide
Solid
304.19
1g
1291773-90-4
97%
MFCD13481969
Videos
SDS