missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromobenzyl)-2-chlorobenzamide, 97%

Brand:  Alfa Aesar™ H59365.03

Code : NEW

Additional Details : CAS Number : 1283105-08-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15405658

  • £358.49 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromobenzyl)-2-chlorobenzamide
1283105-08-7
324.6
MFCD16386278

Specifications

N-(4-Bromobenzyl)-2-chlorobenzamide
Solid
324.6
1g
1283105-08-7
97%
MFCD16386278
Videos
SDS