missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromobenzyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65778.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15442629

  • £119.78 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromobenzyl)-2,2-diphenylacetamide
380.29
MFCD27975816

Specifications

N-(4-Bromobenzyl)-2,2-diphenylacetamide
95%
MFCD27975816
Solid
380.29
1g
Videos
SDS