missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromo-3-methylphenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59259.03

Code : NEW

Additional Details : CAS Number : 330468-23-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15405518

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromo-3-methylphenyl)-3-methylbenzamide
330468-23-0
304.19
MFCD01213549

Specifications

N-(4-Bromo-3-methylphenyl)-3-methylbenzamide
Solid
304.19
Pale brown
330468-23-0
97%
MFCD01213549
1g
Videos
SDS