missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromo-3-fluorophenyl)-4-chloro-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65442.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409646

  • £51.94 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromo-3-fluorophenyl)-4-chloro-3-sulfamoylbenzamide
407.64
MFCD27975763

Specifications

N-(4-Bromo-3-fluorophenyl)-4-chloro-3-sulfamoylbenzamide
95%
MFCD27975763
Solid
407.64
1g
Videos
SDS