Alfa Aesar™ N-(4-Bromo-3-fluorophenyl)-3,5-difluorobenzamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H61419.MD

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15416506

Quantity Price
1 £ 14.95 / 250mg
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-(4-Bromo-3-fluorophenyl)-3,5-difluorobenzamide
Formula Weight 330.1
MDL Number MFCD21526222

Specification

Formula Weight 330.1
Chemical Name or Material N-(4-Bromo-3-fluorophenyl)-3,5-difluorobenzamide
Physical Form Solid
MDL Number MFCD21526222
Assay 97%
Quantity 250mg
Documentation

Technical documents