missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromo-2-methylphenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59710.03

Code : NEW

Additional Details : CAS Number : 349397-95-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15436108

  • £358.49 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromo-2-methylphenyl)-3-methylbenzamide
349397-95-1
304.19
MFCD01213548

Specifications

N-(4-Bromo-2-methylphenyl)-3-methylbenzamide
Solid
304.19
Off-white
349397-95-1
97%
MFCD01213548
1g
Videos
SDS