missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromo-2-methylphenyl)-3,4-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61786.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15466776

  • £56.29 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromo-2-methylphenyl)-3,4-difluorobenzenesulfonamide
362.19
MFCD05116755

Specifications

N-(4-Bromo-2-methylphenyl)-3,4-difluorobenzenesulfonamide
97%
MFCD05116755
Solid
362.19
1g
Videos
SDS