missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromo-2-methylphenyl)-2-chlorobenzamide, 97%

Brand:  Alfa Aesar™ H59162.03

Code : NEW

Additional Details : CAS Number : 327991-20-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475388

  • £358.49 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromo-2-methylphenyl)-2-chlorobenzamide
327991-20-8
324.6
MFCD00739373

Specifications

N-(4-Bromo-2-methylphenyl)-2-chlorobenzamide
Solid
324.6
1g
327991-20-8
97%
MFCD00739373
Videos
SDS