missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromo-2-fluorophenyl)-2-fluorobenzamide, 97%

Brand:  Alfa Aesar™ H59489.03

Code : NEW

Additional Details : CAS Number : 551906-53-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15455828

  • £358.49 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromo-2-fluorophenyl)-2-fluorobenzamide
551906-53-7
312.11
MFCD01353404

Specifications

N-(4-Bromo-2-fluorophenyl)-2-fluorobenzamide
Solid
312.11
1g
551906-53-7
97%
MFCD01353404
Videos
SDS