missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromo-2-chlorophenyl)-2-fluorobenzamide, 97%

Brand:  Alfa Aesar™ H59414.03

Code : NEW

Additional Details : CAS Number : 902093-29-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15425728

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromo-2-chlorophenyl)-2-fluorobenzamide
902093-29-2
328.57
MFCD04805162

Specifications

N-(4-Bromo-2-chlorophenyl)-2-fluorobenzamide
Solid
328.57
1g
902093-29-2
97%
MFCD04805162
Videos
SDS