missing translation for 'onlineSavingsMsg'
Learn More

N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulfonamide, 99%, Thermo Scientific Chemicals

Catalog Number 15465505
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
147118-37-4
C16H18FN3O3S
351.396
MFCD08458342
WOCOTUDOVSLFOB-UHFFFAOYSA-N
n-4-4-fluorophenyl-5-formyl-6-isopropylpyrimidin-2-yl-n-methylmethanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonyl amino pyrimidinyl-5-yl-formyl, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino-5-pyrimidinecarboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino pyrimidin-5-carboxaldehyde, n-4-4-fluorophenyl-5-formyl-6-isopropyl-pyrimidin-2-yl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-1-methylethyl-2-pyrimidinyl-n-methyl-methanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carbaldehyde
10473133
N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C
This item is not returnable. View return policy
147118-37-4
C16H18FN3O3S
351.396
MFCD08458342
WOCOTUDOVSLFOB-UHFFFAOYSA-N
n-4-4-fluorophenyl-5-formyl-6-isopropylpyrimidin-2-yl-n-methylmethanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonyl amino pyrimidinyl-5-yl-formyl, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino-5-pyrimidinecarboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino pyrimidin-5-carboxaldehyde, n-4-4-fluorophenyl-5-formyl-6-isopropyl-pyrimidin-2-yl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-1-methylethyl-2-pyrimidinyl-n-methyl-methanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carbaldehyde
10473133
N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 5 g
Formula Weight 351.4
Percent Purity 99%
Physical Form Solid
Chemical Name or Material N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulfonamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'