missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N-[4-(2-Thiazolidinyl)phenyl]acetamide, 97%
Brand: Alfa Aesar™ H62010.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
N-[4-(2-Thiazolidinyl)phenyl]acetamide | |
222.31 | |
MFCD09027147 | |
2-(4-Acetamidophenyl)thiazolidine |
Specifications
N-[4-(2-Thiazolidinyl)phenyl]acetamide | |
Solid | |
222.31 | |
2-(4-Acetamidophenyl)thiazolidine |
1g | |
97% | |
MFCD09027147 |