missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Methylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60496.03

Code : NEW

Additional Details : CAS Number : 13587-57-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15426968

  • £358.49 / 1g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Methylphenyl)benzenesulfonamide
13587-57-0
247.31
MFCD00584621

Specifications

N-(3-Methylphenyl)benzenesulfonamide
1g
Solid
247.31
Pale pink
13587-57-0
97%
MFCD00584621
Videos
SDS