missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N-(3-Isopropylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%
Brand: Alfa Aesar™ H65983.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
N-(3-Isopropylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide | |
293.41 | |
MFCD28098850 |
Specifications
N-(3-Isopropylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide | |
Solid | |
293.41 |
1g | |
95% | |
MFCD28098850 |