missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Iodophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60839.03

Code : NEW

Additional Details : CAS Number : 151721-40-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15437338

  • £358.49 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Iodophenyl)benzenesulfonamide
359.18
9907287
151721-40-3
MFCD01986021

Specifications

N-(3-Iodophenyl)benzenesulfonamide
Pink
151721-40-3
97%
MFCD01986021
Light Sensitive
1g
Solid
359.18
9907287
Videos
SDS