missing translation for 'onlineSavingsMsg'
Learn More

n-(3-Formylphenyl)methanesulfonamide, ≥97%, Maybridge

£54.00 - £92.00

Chemical Identifiers

CAS 55512-05-5
Molecular Formula C8H9NO3S
Molecular Weight (g/mol) 199.224
MDL Number MFCD03198179
InChI Key CBDSSTWZEANOCR-UHFFFAOYSA-N
Synonym n-3-formylphenyl methanesulfonamide, 3-methylsulfonamido benzaldehyde, methanesulfonamide, n-3-formylphenyl, 3-methylsulfonyl amino benzaldehyde, n-3-formylphenyl methanesulphonamide, m-methylsulfonylamino benzaldehyde, 3-methylsulfonylamino benzaldehyde, methanesulfonamide,n-3-formylphenyl, n-3-formyl-phenyl-methanesulfonamide
PubChem CID 2794787
IUPAC Name N-(3-formylphenyl)methanesulfonamide
SMILES CS(=O)(=O)NC1=CC=CC(=C1)C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10147252 SDS View Documents Maybridge
MO07105CB
250mg Amber Glass Bottle £54.00
250mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 29-01-2020
Please to purchase this item. Need a web account? Register with us today!
10004982 SDS View Documents Maybridge
MO07105DA
1g Amber Glass Bottle £92.00
1g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 02-03-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

55512-05-5
199.224
CBDSSTWZEANOCR-UHFFFAOYSA-N
2794787
CS(=O)(=O)NC1=CC=CC(=C1)C=O
C8H9NO3S
MFCD03198179
n-3-formylphenyl methanesulfonamide, 3-methylsulfonamido benzaldehyde, methanesulfonamide, n-3-formylphenyl, 3-methylsulfonyl amino benzaldehyde, n-3-formylphenyl methanesulphonamide, m-methylsulfonylamino benzaldehyde, 3-methylsulfonylamino benzaldehyde, methanesulfonamide,n-3-formylphenyl, n-3-formyl-phenyl-methanesulfonamide
N-(3-formylphenyl)methanesulfonamide

Specifications

N-(3-formylphenyl)methanesulfonamide
250mg
97.0
C8H9NO3S
n-3-formylphenyl methanesulfonamide, 3-methylsulfonamido benzaldehyde, methanesulfonamide, n-3-formylphenyl, 3-methylsulfonyl amino benzaldehyde, n-3-formylphenyl methanesulphonamide, m-methylsulfonylamino benzaldehyde, 3-methylsulfonylamino benzaldehyde, methanesulfonamide,n-3-formylphenyl, n-3-formyl-phenyl-methanesulfonamide
CS(=O)(=O)NC1=CC=CC(=C1)C=O
199.224
199.23
Amber Glass Bottle
55512-05-5
97%
MFCD03198179
CBDSSTWZEANOCR-UHFFFAOYSA-N
N-(3-formylphenyl)methanesulfonamide
2794787
97%