missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Fluorophenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65849.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15462649

  • £132.50 / 1g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Fluorophenyl)-2,2-diphenylacetamide
305.35
MFCD00399736

Specifications

N-(3-Fluorophenyl)-2,2-diphenylacetamide
95%
MFCD00399736
Solid
305.35
1g
Videos
SDS