missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Fluoro-2-methylphenyl)-2-methoxybenzamide, 97%

Brand:  Alfa Aesar™ H59569.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415106

  • £358.49 / 1g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Fluoro-2-methylphenyl)-2-methoxybenzamide
259.28
MFCD22683078

Specifications

N-(3-Fluoro-2-methylphenyl)-2-methoxybenzamide
97%
MFCD22683078
Solid
259.28
1g
Videos
SDS