missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Ethylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60847.03

Code : NEW

Additional Details : CAS Number : 543703-96-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15417348

  • £358.49 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Ethylphenyl)benzenesulfonamide
261.34
15767000
543703-96-4
MFCD03374403

Specifications

N-(3-Ethylphenyl)benzenesulfonamide
Solid
261.34
15767000
1g
543703-96-4
97%
MFCD03374403
Orange
Videos
SDS