missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chlorophenyl)-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62679.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15497716

  • £367.82 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chlorophenyl)-3,5-difluorobenzenesulfonamide
C12H8ClF2NO2S
303.71
MFCD11729688

Specifications

N-(3-Chlorophenyl)-3,5-difluorobenzenesulfonamide
C12H8ClF2NO2S
MFCD11729688
97%
303.71
5g
Videos
SDS