missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(3-Chlorophenyl)-3,5-difluorobenzamide, 97%

Brand:  Alfa Aesar™ H61999.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15418598

  • £63.81 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(3-Chlorophenyl)-3,5-difluorobenzamide
267.66
MFCD04806291

Specifications

N-(3-Chlorophenyl)-3,5-difluorobenzamide
97%
MFCD04806291
Solid
267.66
1g
Videos
SDS